Orders LinkContactUsLinkComponent
Base materials
Peptone from meat, 1 kg
Catalogus # SERA48620.02
Base materials
Catalogus # SERA48620.02
Leverancier:  
CAS-nummer:  
Base materials
Peptone from meat, 1 kg
CAS-nummer:  
Discontinued
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specificaties
Specificaties
  • VPE:
    1 kg
  • Omschrijving:
    Peptone from meat
Specificaties