Orders LinkContactUsLinkComponent
Base materials
Base materials
Peptone from casein (enzymatic digest), Glass bottle for solids, 1 kg
Catalogus # SERA48610.05
Marketsource
Leverancier:   Serva
Base materials
Catalogus # SERA48610.05
Leverancier:  Serva
Marketsource
CAS-nummer:  
Base materials
Peptone from casein (enzymatic digest), Glass bottle for solids, 1 kg
CAS-nummer:  
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 kg
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specificaties
Specificaties
  • Verpakking:
    Glass bottle for solids
  • VPE:
    1 kg
  • Omschrijving:
    Peptone from casein (enzymatic digest)
Specificaties