Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

D-(+)-Sucrose

5 kg
Catalogus # SERA35579.03
Marketsource
Leverancier:   Serva
D-(+)-Sucrose
Catalogus # SERA35579.03
Leverancier:  Serva
Marketsource
CAS-nummer:  57-50-1
D-(+)-Sucrose
D-(+)-Sucrose

5 kg

CAS-nummer:  57-50-1
Price
136,00 €
Log in voor beschikbaarheid
1 * 5 kg
Product Details & Documents
Synoniemen:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS-nummer:57-50-1
  • Formule:C₁₂H₂₂O₁₁
  • MDL-nummer:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck index:13,08966
  • Smelten Pt:169…170 °C
  • Dichtheid:1,29 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:200-334-9
Specificaties

Specificaties

  • VPE:
    5 kg

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.