Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Sucrose

D-(+)-Sucrose

Glass bottle for solids, 500 g
Catalogus # SERA35579.02
Marketsource
Leverancier:   Serva
D-(+)-Sucrose
Catalogus # SERA35579.02
Leverancier:  Serva
Marketsource
CAS-nummer:  57-50-1
D-(+)-Sucrose
D-(+)-Sucrose

Glass bottle for solids, 500 g

CAS-nummer:  57-50-1
Price
23,00 €
Log in voor beschikbaarheid
1 * 500 g
Product Details & Documents
Synoniemen:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS-nummer:57-50-1
  • Formule:C₁₂H₂₂O₁₁
  • MDL-nummer:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck index:13,08966
  • Smelten Pt:169…170 °C
  • Dichtheid:1,29 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:200-334-9
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    500 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.