D-(+)-Sucrose
Glass bottle for solids, 100 g
Catalogus # SERA35579.01
Marketsource
Leverancier: Serva
Ga naar:
Product Details & Documents
Synoniemen: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
- CAS-nummer:57-50-1
- Formule:C₁₂H₂₂O₁₁
- MDL-nummer:MFCD00006626
- Molecular Weight:342.3 g/mol
- Merck index:13,08966
- Smelten Pt:169…170 °C
- Dichtheid:1,29 g/cm³ (20 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:200-334-9
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:100 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



