Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Phthalaldehyde

Phthalaldehyde

Glass bottle for solids, 5 g
Catalogus # SERA32800.01
Giftig
Corrosief
Milieu
Marketsource
Leverancier:   Serva
Phthalaldehyde
Catalogus # SERA32800.01
Leverancier:  Serva
Marketsource
CAS-nummer:  643-79-8
Phthalaldehyde

Glass bottle for solids, 5 g

CAS-nummer:  643-79-8
Price
38,00 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
Synoniemen:  1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
  • CAS-nummer:643-79-8
  • Formule:C₈H₆O₂
  • MDL-nummer:MFCD00003335
  • Molecular Weight:134.13 g/mol
  • Kokend Pt:266 °C (1013 hPa)
  • Smelten Pt:54…56 °C
  • Dichtheid:1,13 g/cm³ (20 °C)
  • Flits Pt:132 °C
  • EINECS:211-402-2
  • UN:2923
  • ADR:8,II
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5 g

Specificaties