Orders LinkContactUsLinkComponent
Specimen loops
Specimen loop, 0,5 mm, 1 mm, 40 mm
Catalog # 391-0134
Supplier:   KLS Martin Group
Specimen loops
Catalog # 391-0134
Supplier:  KLS Martin Group
Specimen loops
Specimen loop, 0,5 mm, 1 mm, 40 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Ø:
    1 mm
  • Omschrijving:
    Specimen loop, 0,5 mm
  • Lengte:
    40 mm