Orders LinkContactUsLinkComponent
Büchner trechters, DURAN®
Büchner trechters, DURAN®
120 mm, 1000 ml, 110 mm
Catalog # 511-0339
Supplier:   DWK Life Sciences
Büchner trechters, DURAN®
Catalog # 511-0339
Supplier:  DWK Life Sciences
Büchner trechters, DURAN®
Büchner trechters, DURAN®
120 mm, 1000 ml, 110 mm
Price
290,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicaatglas 3.3, helder.
For filter papers or membrane filtration discs.
Specifications
  • Ø plaat:
    120 mm
  • Capaciteit:
    1000 ml
  • Voor filter Ø:
    110 mm