Vacuum filter holders
130 ml, 47 mm
Catalog # SART16523
Supplier: Sartorius
Jump to:
Product Details & Documents
These vacuum filter holders feature a glass cylinder, PP base and sealing plug with anodised aluminum closing cap.
- Vacuum operated
Specifications
- Voor Ø:47 mm
- Capaciteit:130 ml
- Seq:02
Accessories
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice