Biology prepared microscope slides, school set B
Fibrous connective tissue of mammal
Catalog # LIED618E
Supplier: LIEDER JOHANNES
Jump to:
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
: Supplied on high quality ground edge slides (26×76 mm).
Specifications
- Omschrijving:Fibrous connective tissue of mammal
- Seq:210