Biology prepared microscope slides, school set B
Paramecium stained nuclei
LIED601D
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
: Supplied on high quality ground edge slides (26×76 mm).
- Omschrijving:Paramecium stained nuclei
- Seq:202