Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Biology prepared microscope slides, school set B
Paramecium stained nuclei
  LIED601D
Biology prepared microscope slides, school set B
  LIED601D
Biology prepared microscope slides, school set B
Biology prepared microscope slides, school set B
Paramecium stained nuclei
8,90 €
1 * 1 ST
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
 : Supplied on high quality ground edge slides (26×76 mm).
  • Omschrijving:
    Paramecium stained nuclei
  • Seq:
    202