Invertebrates prepared slides
Amoeba proteus nucleus and pseudopodia
Catalogus # LIED2101E
Leverancier: LIEDER JOHANNES
Ga naar:
Product Details & Documents
This set of invertebrates includes representative specimen of protozoa, sponges, coelenterata, worms, arthropoda, insects, molluscs, echinoderms and Acraniata.
Livraison: Supplied with accompanying brochure.
Specificaties
- Omschrijving:Amoeba proteus nucleus and pseudopodia