Orders LinkContactUsLinkComponent
Adapters, 3 halzen
Adapters, 3 halzen
29/32 NS, 29/32 NS
Catalogus # 201-2221
Leverancier:   Lenz Laborglas
Adapters, 3 halzen
Catalogus # 201-2221
Leverancier:  Lenz Laborglas
Adapters, 3 halzen
Adapters, 3 halzen
29/32 NS, 29/32 NS
Price
91,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
DURAN® Borosilicate glass 3.3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 3 NS sockets and 1 NS cone, with two parallel necks and one angled neck (45°).
Documents
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Huls:
    29/32 NS