NS-stoppen
19/26, Met afdruiptip
217-9106
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
- Excellent chemical resistance
- High thermal resistance, minimal heat expansion results in high resistance to temperature changes
- Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
: DIN 12252
: Available with flat or pointed bottom.
- (NS):19/26
- Type:Met afdruiptip
- Seq:05