Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
NS-stoppen
19/26, Met afdruiptip
  217-9106
NS-stoppen
  217-9106
NS-stoppen
NS-stoppen
19/26, Met afdruiptip
4,10 €
1 * 1 ST
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
 : DIN 12252
 : Available with flat or pointed bottom.
  • (NS):
    19/26
  • Type:
    Met afdruiptip
  • Seq:
    05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.