NS-stoppen
14/23, Met afdruiptip
Catalogus # 217-9105
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
- Excellent chemical resistance
- High thermal resistance, minimal heat expansion results in high resistance to temperature changes
- Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certificeringen: DIN 12252
Bestelinformatie: Available with flat or pointed bottom.
Documents
BSE TSE Glass Columns
BSE/TSE-certificaten
Specificaties
- (NS):14/23
- Type:Met afdruiptip
- Seq:04