Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
NS-stoppen
NS-stoppen
14/23, Met afdruiptip
Catalogus # 217-9105
Leverancier:   Lenz Laborglas
NS-stoppen
Catalogus # 217-9105
Leverancier:  Lenz Laborglas
NS-stoppen
NS-stoppen
14/23, Met afdruiptip
Price
3,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certificeringen: DIN 12252
Bestelinformatie: Available with flat or pointed bottom.
Documents
Specificaties
  • (NS):
    14/23
  • Type:
    Met afdruiptip
  • Seq:
    04