Weegpotjes, PP
50 ml, 59×34 mm
Catalog # 611-9404
Supplier: KARTELL
Jump to:
Product Details & Documents
PP met deksel.
- Very good chemical resistance
- Very good temperature resistance
Cylindrical shape, with knobbed lid.
Documents
Specifications
- Ø×h:59×34 mm
- Capaciteit:50 ml