Weegpotjes, PP
60 ml, 40×69 mm
Catalogus # 611-0104
Leverancier: KARTELL
Ga naar:
Product Details & Documents
PP met deksel.
- Very good chemical resistance
- Very good temperature resistance
Cylindrical shape, with knobbed lid.
Specificaties
- Ø×h:40×69 mm
- Capaciteit:60 ml