Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Roerstaven

Roerstaven

Roerstaaf, Glas, 6 - 7 mm, 250 mm
Catalog # 441-0263
Supplier:   EMMANUEL PIERRE
Roerstaven
Catalog # 441-0263
Supplier:  EMMANUEL PIERRE
Roerstaven
Roerstaven

Roerstaaf, Glas, 6 - 7 mm, 250 mm

Price
Inquire For Price
Log in for availability
1 * 10 ST
Product Details & Documents

Uit gewoon glas.

Specifications
  • Ø:
    6 - 7 mm
  • Omschrijving:
    Roerstaaf
  • Lengte:
    250 mm
  • Materiaal:
    Glas
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.