2-Ethylimidazool 99%
Glass bottle for solids, 100 g
: 2-Ethyl-1H-imidazole
- :1072-62-4
- :C₅H₈N₂
- :MFCD00005192
- :96.13 g/mol
- :268 °C (760 mmHg)
- :83…87 °C
- :1,03 g/cm³ (25 °C)
- :159 °C
- :Kamertemperatuur
- :214-011-5
- Appearance:White to light yellow Flakes
- Infrared spectrum:Conforms
- Melting point:83 to 87 °C
- Water:≤0.5 %
- GC:≥98.5 %
- Verpakking:Glass bottle for solids
- VPE:100 g



