Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Kwikdichloride 99.5%

Kwikdichloride 99.5%

Plastic bottle, 100 g
Catalogus # ACRO201431000
Giftig
Gezondheid
Corrosief
Milieu
Leverancier:   Thermo Fisher Scientific
Kwikdichloride 99.5%
Catalogus # ACRO201431000
CAS-nummer:  7487-94-7
Kwikdichloride 99.5%
Kwikdichloride 99.5%

Plastic bottle, 100 g

CAS-nummer:  7487-94-7
Price
62,80 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents
Synoniemen:  Mercuric chloride, Mercury Chloride, Mercury dichloride
  • CAS-nummer:7487-94-7
  • Formule:HgCl₂
  • MDL-nummer:MFCD00011041
  • Molecular Weight:271.5 g/mol
  • Merck index:13,05901
  • Kokend Pt:302 °C (760 mmHg)
  • Smelten Pt:277 °C
  • Dichtheid:5,44 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:231-299-8
  • UN:1624
  • ADR:6.1,II

Documents

Specificaties

Specificatie testresultaten

  • Appearance:
    White Crystalline powder or crystals (May contain black particles)
  • Titration Complexometric:
    ≥99.4 %

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    100 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.