Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Sulphamide 98+%

Sulphamide 98+%

Glass bottle, 25 g
Catalogus # ACRO199020250
Irriterend
Leverancier:   Thermo Fisher Scientific
Sulphamide 98+%
Catalogus # ACRO199020250
CAS-nummer:  7803-58-9
Sulphamide 98+%
Sulphamide 98+%

Glass bottle, 25 g

CAS-nummer:  7803-58-9
Price
197,00 €
Log in voor beschikbaarheid
1 * 25 g
Product Details & Documents
Synoniemen:  Sulphuric diamide
  • CAS-nummer:7803-58-9
  • Formule:H₄N₂O₂S
  • MDL-nummer:MFCD00011606
  • Molecular Weight:96.11 g/mol
  • Smelten Pt:87…91 °C
  • Dichtheid:1,61 g/cm³
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:232-262-9

Documents

Specificaties

Specificatie testresultaten

  • Appearance:
    White to practically white Crystalline powder (Free from visible impurities)
  • Infrared spectrum:
    Conforms
  • Melting point:
    89.0 to 93.0 °C
  • Loss on drying:
    ≤0.5 % (50 °C, 2 hrs) (vacuum)
  • Elemental Analysis:
    ≥98.0 %
  • Related substances:
    (TLC) Passes test
  • pH:
    5.0 to 7.0 (10 % in water)

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    25 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.