Natriumazide 99%, extra pure
Plastic bottle, 100 g
: Sodium triazide
- :26628-22-8
- :NaN₃
- :MFCD00003536
- :65.01 g/mol
- :13,08653
- :275 °C
- :1,85 g/cm³
- :Kamertemperatuur
- :247-852-1
- :1687
- :6.1,II
- Appearance:White Crystalline powder
- Assay:≥98.5 %
- Loss on drying:≤0.5 %
- pH:≥9 (5 % solution)
- Sodium carbonate (Na2CO3):≤0.15 %
- Sodium hydroxide (NaOH):≤0.1 %
- Heavy metals (as Pb):≤20 ppm
- Insoluble matter:≤0.05 % (in water)
- Verpakking:Plastic bottle
- VPE:100 g



