Natriumazide 99%, extra pure
Glass bottle for solids, 5 g
Leverancier: Thermo Fisher Scientific
Natriumazide 99%, extra pure
Catalogus # ACRO190380050
Leverancier: Thermo Fisher Scientific
CAS-nummer: 26628-22-8
Ga naar:
Product Details & Documents
Synoniemen: Sodium triazide
- CAS-nummer:26628-22-8
- Formule:NaN₃
- MDL-nummer:MFCD00003536
- Molecular Weight:65.01 g/mol
- Merck index:13,08653
- Smelten Pt:275 °C
- Dichtheid:1,85 g/cm³
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
Documents
Certificaten
Zoeken op lotnummerACRO19038
SDS
Thermo Scientific Stockroom Essentials
Product documentatie
Specificaties
Specificatie testresultaten
- Appearance:White Crystalline powder
- Assay:≥98.5 %
- Loss on drying:≤0.5 %
- pH:≥9 (5 % solution)
- Sodium carbonate (Na2CO3):≤0.15 %
- Sodium hydroxide (NaOH):≤0.1 %
- Heavy metals (as Pb):≤20 ppm
- Insoluble matter:≤0.05 % (in water)
Specificaties
- Verpakking:Glass bottle for solids
- VPE:5 g



