Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Celcultuurinserts, Falcon®

Celcultuurinserts, Falcon®

Inserts for 6-well plates, sterile, Transparent PET membrane, 3,0 μm
Catalogus # 734-0033
Leverancier:   Corning
Celcultuurinserts, Falcon®
Catalogus # 734-0033
Leverancier:  Corning
Celcultuurinserts, Falcon®
Celcultuurinserts, Falcon®

Inserts for 6-well plates, sterile, Transparent PET membrane, 3,0 μm

Price
468,64 €
Log in voor beschikbaarheid
1 * 48 ST
Product Details & Documents

Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.

  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12 and 24 well
  • A broad selection of membrane pore sizes, 0,4, 1,0, 3,0 and 8,0 µm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture
Specificaties
  • Membraan:
    Transparent PET membrane
  • Poriëngrootte:
    3,0 μm
  • Omschrijving:
    Inserts for 6-well plates, sterile
  • Seq:
    10