Cellulose triacetate
Plastic bottle, 250 g
Leverancier: Thermo Fisher Scientific
Cellulose triacetate
Catalogus # ACRO177822500
Leverancier: Thermo Fisher Scientific
CAS-nummer: 9012-09-3
Ga naar:
Product Details & Documents
Appearance: White to light yellow Pellets or rods
- CAS-nummer:9012-09-3
- Formule:C₄₀H₅₄O₂₇
- Molecular Weight:966.845 g/mol
- Temperatuur bij opslag:Kamertemperatuur
Documents
ACRO17782
SDS
Specificaties
Specificatie testresultaten
- Appearance:White to light yellow Pellets or rods
- Infrared spectrum:Conforms
- Acetyl content:43 to 44 wt %
- Titration Argentometric:≥95 % (on dry substance)
- Water:≤10 %
Specificaties
- Verpakking:Plastic bottle
- VPE:250 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



