Testcapsule
TX23L, test cap, Lumberg type, 0 °C
Catalogus # 620-0377
Leverancier: COMARK
Ga naar:
Product Details & Documents
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.
Specificaties
- Meetbereik:0 °C
- Omschrijving:TX23L, test cap, Lumberg type