Natriumhydroxide 98%, pellets, extra pure
Plastic bottle, 2,5 kg
Leverancier: Thermo Fisher Scientific
Natriumhydroxide 98%, pellets, extra pure
Catalogus # ACRO134070025
Leverancier: Thermo Fisher Scientific
CAS-nummer: 1310-73-2
Ga naar:
Product Details & Documents
Synoniemen: Caustic soda
- CAS-nummer:1310-73-2
- Formule:NaOH
- MDL-nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck index:14,08627
- Kokend Pt:1390 °C (760 mmHg)
- Smelten Pt:318 °C
- Dichtheid:2,13 g/cm³ (20 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- BEREIKEN:01-2119457892-27
Documents
ACRO13407
SDS
Specificaties
Specificatie testresultaten
- Appearance:White Pellets
- Titration with HCl:≥97.5 %
- Chloride (Cl):≤50 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤40 ppm
- Sodium carbonate (Na2CO3):≤1.0 %
- Aluminium (Al):≤10 ppm
- Calcium (Ca):≤10 ppm
- Iron (Fe):≤5 ppm
- Lead (Pb):≤1 ppm
- Nickel (Ni):≤5 ppm
- Potassium (K):≤250 ppm
- Silver (Ag):≤0.5 ppm
- Silicon dioxide (SiO2):≤5 ppm
- Nitrogen compounds (as N):≤0.0005 %
Specificaties
- Verpakking:Plastic bottle
- VPE:2,5 kg
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



