Natriumhydroxide 98%, pellets, extra pure
Plastic bottle for solids, 1 kg
Supplier: Thermo Fisher Scientific
Natriumhydroxide 98%, pellets, extra pure
Catalog # ACRO134070010
Supplier: Thermo Fisher Scientific
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (760 mmHg)
- Melting Pt:318 °C
- Density:2,13 g/cm³ (20 °C)
- Storage Temperature:Kamertemperatuur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
ACRO13407
SDS
Specifications
Specification Test Results
- Appearance:White Pellets
- Titration with HCl:≥97.5 %
- Chloride (Cl):≤50 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤40 ppm
- Sodium carbonate (Na2CO3):≤1.0 %
- Aluminium (Al):≤10 ppm
- Calcium (Ca):≤10 ppm
- Iron (Fe):≤5 ppm
- Lead (Pb):≤1 ppm
- Nickel (Ni):≤5 ppm
- Potassium (K):≤250 ppm
- Silver (Ag):≤0.5 ppm
- Silicon dioxide (SiO2):≤5 ppm
- Nitrogen compounds (as N):≤0.0005 %
Specifications
- Verpakking:Plastic bottle for solids
- VPE:1 kg



