Styreen 99% stabilised, extra pure
Glass bottle, 2,5 L
Leverancier: Thermo Fisher Scientific
Styreen 99% stabilised, extra pure
Catalogus # ACRO132790025
Leverancier: Thermo Fisher Scientific
CAS-nummer: 100-42-5
Ga naar:
Product Details & Documents
Synoniemen: Phenylethene, Phenylethylene, Vinylbenzene
- CAS-nummer:100-42-5
- Formule:C₈H₈
- MDL-nummer:MFCD00008612
- Molecular Weight:104.15 g/mol
- Merck index:13,08944
- Kokend Pt:145…146 °C (760 mmHg)
- Smelten Pt:–31 °C
- Dichtheid:0,906 g/cm³
- Flits Pt:31 °C
- Temperatuur bij opslag:Koelkast
- EINECS:202-851-5
- UN:2055
- ADR:3,III
Documents
Specificaties
Specificatie testresultaten
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5460 to 1.5480 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Stabilizer:10 to 20 ppm p-tert-Butylcatechol
- Peroxides:≤0.01 % (as H2O2)
Specificaties
- Verpakking:Glass bottle
- VPE:2,5 L



