Styreen 99% stabilised, extra pure
Glass bottle, 1 L
Leverancier: Thermo Fisher Scientific
Styreen 99% stabilised, extra pure
Catalogus # ACRO132790010
Leverancier: Thermo Fisher Scientific
CAS-nummer: 100-42-5
Ga naar:
Product Details & Documents
Synoniemen: Phenylethene, Phenylethylene, Vinylbenzene
- CAS-nummer:100-42-5
- Formule:C₈H₈
- MDL-nummer:MFCD00008612
- Molecular Weight:104.15 g/mol
- Merck index:13,08944
- Kokend Pt:145…146 °C (760 mmHg)
- Smelten Pt:–31 °C
- Dichtheid:0,906 g/cm³
- Flits Pt:31 °C
- Temperatuur bij opslag:Koelkast
- EINECS:202-851-5
- UN:2055
- ADR:3,III
Documents
Specificaties
Specificatie testresultaten
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5460 to 1.5480 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Stabilizer:10 to 20 ppm p-tert-Butylcatechol
- Peroxides:≤0.01 % (as H2O2)
Specificaties
- Verpakking:Glass bottle
- VPE:1 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



