Phthalaldehyde 98+%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
- CAS Number:643-79-8
- Formula:C₈H₆O₂
- MDL Number:MFCD00003335
- Molecular Weight:134.13 g/mol
- Boiling Pt:83…84 °C (1hPa)
- Melting Pt:54…57 °C
- Density:1,13 g/cm³ (20 °C)
- Flash Pt:132 °C
- Storage Temperature:Koelkast
- EINECS:211-402-2
- UN:2923
- ADR:8,II
Documents
Specifications
Specification Test Results
- Appearance:Yellow Powder or crystals and/or lumps
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥98.0 %
Specifications
- Verpakking:Glass bottle for solids
- VPE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



