Maleïnezuur 99%
Plastic bottle, 100 g
- :110-16-7
- :C₄H₄O₄
- :MFCD00063177
- :116.07 g/mol
- :13,05726
- :256 °C (1013 hPa)
- :134…138 °C
- :1.59 g/cm³ (20 °C)
- :127 °C
- :Kamertemperatuur
- :203-742-5
- :3261
- :8,III
- Appearance:White Crystalline powder or granules and/or chunks
- Infrared spectrum:Conforms
- Melting point:130 to 138 °C
- Water:≤2 %
- HPLC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 % (on dry substance)
- Heavy metals (as Pb):≤10 ppm
- Verpakking:Plastic bottle
- VPE:100 g



