Maleïnezuur 99%
Plastic bottle, 1 kg
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
- CAS-nummer:110-16-7
- Formule:C₄H₄O₄
- MDL-nummer:MFCD00063177
- Molecular Weight:116.07 g/mol
- Merck index:13,05726
- Kokend Pt:256 °C (1013 hPa)
- Smelten Pt:134…138 °C
- Dichtheid:1.59 g/cm³ (20 °C)
- Flits Pt:127 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:203-742-5
- UN:3261
- ADR:8,III
Documents
ACRO12523
SDS
Specificaties
Specificatie testresultaten
- Appearance:White Crystalline powder or granules and/or chunks
- Infrared spectrum:Conforms
- Melting point:130 to 138 °C
- Water:≤2 %
- HPLC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 % (on dry substance)
- Heavy metals (as Pb):≤10 ppm
Specificaties
- Verpakking:Plastic bottle
- VPE:1 kg
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



