Ethyleendiamine 99%
Glass bottle, 2,5 L
Leverancier: Thermo Fisher Scientific
Ethyleendiamine 99%
Catalogus # ACRO118420025
Leverancier: Thermo Fisher Scientific
CAS-nummer: 107-15-3
Ga naar:
Product Details & Documents
Synoniemen: 1,2-Ethanediamine, 1,2-Diaminoethane
- CAS-nummer:107-15-3
- Formule:C₂H₈N₂
- MDL-nummer:MFCD00008204
- Molecular Weight:60.1 g/mol
- Merck index:13,03829
- Kokend Pt:117…118 °C (760 mmHg)
- Smelten Pt:11 °C
- Dichtheid:0,898 g/cm³
- Flits Pt:38 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:203-468-6
- UN:1604
- ADR:8,II
Specificaties
Specificatie testresultaten
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4555 to 1.4575 (20°C, 589 nm)
- Water:≤0.5 % (Coulometric)
- GC:≥98.5 %
- Sulfate (SO4):≤0.02 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤0.002 %
- Salicylic acid:≤0.04 % (HOC6H4COOH)
- Insoluble matter:≤0.02 %
Specificaties
- Verpakking:Glass bottle
- VPE:2,5 L



