Methyleenchloride 99.8+% stabilised for analysis
Glass bottle, 2,5 L
: Methylene chloride, DCM
- :75-09-2
- :CH₂Cl₂
- :MFCD00000881
- :84.93 g/mol
- :12,06140
- :39,8 °C (1013 hPa)
- :–97 °C
- :1,325 g/cm³
- :Kamertemperatuur
- :200-838-9
- :6.1,III
- :Stabilised with amylene
- :1593
- Appearance:Clear liquid
- Water:≤0.02 % (Coulometric)
- GC:≥99.80 %
- Color scale:≤10 APHA
- Stabilizer:15 to 100 ppm Amylene
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0003 meq/g
- Substance darkened by H2SO4:≤5 APHA
- Free chlorine:≤1 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Verpakking:Glass bottle
- VPE:2,5 L



