Methyleenchloride 99.8+% stabilised for analysis
Glass bottle, 1 L
Leverancier: Thermo Fisher Scientific
Methyleenchloride 99.8+% stabilised for analysis
Catalogus # 113460010.
Leverancier: Thermo Fisher Scientific
CAS-nummer: 75-09-2
Ga naar:
Product Details & Documents
Synoniemen: Methylene chloride, DCM
- CAS-nummer:75-09-2
- Formule:CH₂Cl₂
- MDL-nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck index:12,06140
- Kokend Pt:39,8 °C (1013 hPa)
- Smelten Pt:–97 °C
- Dichtheid:1,325 g/cm³
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with amylene
- UN:1593
Documents
ACRO11346
SDS
Specificaties
Specificatie testresultaten
- Appearance:Clear liquid
- Water:≤0.02 % (Coulometric)
- GC:≥99.80 %
- Color scale:≤10 APHA
- Stabilizer:15 to 100 ppm Amylene
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0003 meq/g
- Substance darkened by H2SO4:≤5 APHA
- Free chlorine:≤1 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
Specificaties
- Verpakking:Glass bottle
- VPE:1 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



