Orders LinkContactUsLinkComponent
Water
Water
Glass bottle, 1 L
Catalog # AMBI9922
Marketsource
Supplier:   Ambion
Water
Catalog # AMBI9922
Supplier:  Ambion
Marketsource
CAS Number:  7732-18-5
Water
Glass bottle, 1 L
CAS Number:  7732-18-5
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:231-791-2
Documents
Specifications
Specifications
  • Verpakking:
    Glass bottle
  • VPE:
    1 L
Specifications