Hydrometers, Brix
60 - 90 °Bx, 0,1 °Bx, 300 mm
Catalog # ALLA3910FC060/20QP
Supplier: Alla France
Jump to:
Product Details & Documents
These Brix sugar scale hydrometers are specially designed for use in breweries.
Specifications
- Lengte:300 mm
- Indeling:0,1 °Bx
- Meetbereik:60 - 90 °Bx