Orders LinkContactUsLinkComponent
Trechters
Trechters
85 mm, 11 mm, 120 mm
Catalog # BRND148237
Supplier:   Brand
Trechters
Catalog # BRND148237
Supplier:  Brand
Trechters
Trechters
85 mm, 11 mm, 120 mm
Price
10,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PP, translucent.
  • External flutings allow air to escape during filtration
  • Excellent chemical resistance, autoclavable
  • Suitable for use with standard filter papers
For rapid filtration.
Specifications
  • Trechter uitw. Ø:
    120 mm
  • Steel uitw. Ø:
    11 mm
  • Lengte steel:
    85 mm