Methylcyclopentane 95%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Methylcyclopentane 95%
Catalog # ACRO345245000
Supplier: Thermo Fisher Scientific
CAS Number: 96-37-7
Jump to:
Product Details & Documents
- CAS Number:96-37-7
- Formula:C₆H₁₂
- MDL Number:MFCD00001382
- Molecular Weight:84.16 g/mol
- Boiling Pt:71 °C
- Melting Pt:–142,4 °C
- Density:0,748 g/cm³
- Flash Pt:–4 °C
- Storage Temperature:Kamertemperatuur
- EINECS:202-503-2
- UN:2298
- ADR:3,II
Documents
ACRO34524
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4070 to 1.4110 (20°C, 589 nm)
- GC:≥94 %
Specifications
- Verpakking:Glass bottle
- VPE:500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



