Orders LinkContactUsLinkComponent
Double bossheads
Double bossheads
Double bosshead, with heart screw M6/M8, 16,5 mm
Catalog # 241-3028
Supplier:   BOCHEM
Double bossheads
Catalog # 241-3028
Supplier:  BOCHEM
Double bossheads
Double bossheads
Double bosshead, with heart screw M6/M8, 16,5 mm
Price
59,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Double bossheads made of stainless steel 18/10.
  • For rods up to 16,5 mm
  • Angle: 90°
  • Available with heart screw M6/M8, socket screw M6/M8 or safety screw type 'heart' M8
Certifications: DIN 12895
Specifications
  • Omschrijving:
    Double bosshead, with heart screw M6/M8
  • Voor staaf Ø:
    16,5 mm