Orders LinkContactUsLinkComponent
Retort rods, with M10 thread
Retort rods, with M10 thread
13 mm, 600 mm, M10
Catalog # BOCH5171
Supplier:   BOCHEM
Retort rods, with M10 thread
Catalog # BOCH5171
Supplier:  BOCHEM
Retort rods, with M10 thread
Retort rods, with M10 thread
13 mm, 600 mm, M10
Price
31,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 18/10.
  • Rods for stand bases
  • M10 thread
Specifications
  • Ø:
    13 mm
  • Schroefdraad:
    M10
  • Lengte:
    600 mm
  • Seq:
    11