N-Methylethylamine 97%
Glass bottle for solids, 5 g
Leverancier: Thermo Fisher Scientific
N-Methylethylamine 97%
Catalogus # ACRO314680050
Leverancier: Thermo Fisher Scientific
CAS-nummer: 624-78-2
Ga naar:
Product Details & Documents
Synoniemen: N-Ethylmethylamine
- CAS-nummer:624-78-2
- Formule:C₃H₉N
- MDL-nummer:MFCD00009030
- Molecular Weight:59.11 g/mol
- Kokend Pt:33…34 °C (760 mmHg)
- Dichtheid:0,68 g/cm³
- Flits Pt:<–34 °C
- Temperatuur bij opslag:Koelkast
- EINECS:210-862-1
- UN:2733
- ADR:3,I
Specificaties
Specificatie testresultaten
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3730 to 1.3770 (20°C, 589 nm)
- GC:≥96.0 %
Specificaties
- Verpakking:Glass bottle for solids
- VPE:5 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



