Pincetten
Spits/gebogen, 130 mm
Catalogus # 232-0019
Leverancier: BOCHEM
Ga naar:
Product Details & Documents
Roestvrij staal, PTFE-gecoat.
- Bent, with guide pin
- Smooth, coated surface to prevent contamination and facilitate cleaning
Specificaties
- Type:Spits/gebogen
- Lengte:130 mm
- Cat. No.:232-0019