Wire baskets, autoclavable, Poxygrid®
360×280×90 mm
Catalogus # BELAH167660000
Leverancier: Bel-Art Products, a Part of SP
Wire baskets, autoclavable, Poxygrid®
Catalogus # BELAH167660000
Leverancier: Bel-Art Products, a Part of SP
Ga naar:
Product Details & Documents
These epoxy-coated, steel wire baskets are suitable for transporting and batch dipping glassware.
- Resistant to rust, corrosion, solvents, acids and alkalis
- Withstands a temperature range of −223 to +121 °C
Specificaties
- b×d×h:360×280×90 mm