L(-)-Mannose 99+%
Glass bottle for solids, 250 mg
Leverancier: Thermo Fisher Scientific
L(-)-Mannose 99+%
Catalogus # ACRO227592500
Leverancier: Thermo Fisher Scientific
CAS-nummer: 10030-80-5
Ga naar:
Product Details & Documents
Synoniemen: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
- CAS-nummer:10030-80-5
- Formule:C₆H₁₂O₆
- MDL-nummer:MFCD00136021
- Molecular Weight:180.16 g/mol
- Kokend Pt:376 °C (1013 hPa)
- Smelten Pt:129…131 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:233-080-2
Documents
ACRO22759
SDS
Specificaties
Specificatie testresultaten
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:-13.5° to -15.5° (20°C, 589 nm) (c=4, water, NH3)
- Heavy metals (as Pb):≤10 ppm
Specificaties
- Verpakking:Glass bottle for solids
- VPE:250 mg
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



