L(-)-Mannose 99+%
Glass bottle for solids, 1 g
Leverancier: Thermo Fisher Scientific
L(-)-Mannose 99+%
Catalogus # ACRO227590010
Leverancier: Thermo Fisher Scientific
CAS-nummer: 10030-80-5
Ga naar:
Product Details & Documents
Synoniemen: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
- CAS-nummer:10030-80-5
- Formule:C₆H₁₂O₆
- MDL-nummer:MFCD00136021
- Molecular Weight:180.16 g/mol
- Kokend Pt:376 °C (1013 hPa)
- Smelten Pt:129…131 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:233-080-2
Specificaties
Specificatie testresultaten
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:-13.5° to -15.5° (20°C, 589 nm) (c=4, water, NH3)
- Heavy metals (as Pb):≤10 ppm
Specificaties
- Verpakking:Glass bottle for solids
- VPE:1 g



