Plakker
EASYseal adhesive film, non sterile, 120×80 mm, Transparent, -
Catalogus # GREI676002_200
Leverancier: Greiner Bio-One
Ga naar:
Product Details & Documents
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
- Free from RNase, DNase and human DNA
- Non pyrogenic
Specificaties
- Optische eigenschap:Transparent
- Steriel:-
- Omschrijving:EASYseal adhesive film, non sterile, 120×80 mm
- Seq:01