Orders LinkContactUsLinkComponent
Sieves
Sieves
Stainless steel sieve, 3,00 mm, Zeef MF 3.0
Catalogus # 412-1135
Leverancier:   IKA
Sieves
Catalogus # 412-1135
Leverancier:  IKA
Sieves
Sieves
Stainless steel sieve, 3,00 mm, Zeef MF 3.0
Price
370,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Specificaties
  • Type:
    Zeef MF 3.0
  • Omschrijving:
    Stainless steel sieve, 3,00 mm
  • Seq:
    10