Orders LinkContactUsLinkComponent
Celtellers, Fuchs-Rosenthal
Celtellers, Fuchs-Rosenthal
Cell counter with clips, bright-lined, 0,2 mm
Catalogus # HIRS8110210
Leverancier:   HIRSCHMANN
Celtellers, Fuchs-Rosenthal
Catalogus # HIRS8110210
Leverancier:  HIRSCHMANN
Celtellers, Fuchs-Rosenthal
Celtellers, Fuchs-Rosenthal
Cell counter with clips, bright-lined, 0,2 mm
Price
185,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Telkamers met dubbele nettotelling.
Bestelinformatie: Includes two cover glasses.
Specificaties
  • Kamerdiepte:
    0,2 mm
  • Omschrijving:
    Cell counter with clips, bright-lined