Celtellers, Fuchs-Rosenthal
Cell counter with clips, bright-lined, 0,2 mm
Catalogus # HIRS8110210
Leverancier: HIRSCHMANN
Ga naar:
Product Details & Documents
Telkamers met dubbele nettotelling.
Bestelinformatie: Includes two cover glasses.
Specificaties
- Kamerdiepte:0,2 mm
- Omschrijving:Cell counter with clips, bright-lined